MAT196007233 Matrix Scientific

N-[(1-Butyl-1,2,3,4-tetrahydroquinolin-6-yl)-methyl]-N-(2-furylmethyl)amine

  • MDL number MFCD12027322
  • Cas Number : 1119453-02-9
  • Molecular Weight : 298.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038426.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038426.html
Availability details for
Quantity
  • Availability
  • Add to Cart