MAT196007241 Matrix Scientific

N-(sec-Butyl)-N-[(1-propyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]amine

  • MDL number MFCD12027328
  • Cas Number : 1119450-28-0
  • Molecular Weight : 260.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038434.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038434.html
Availability details for
Quantity
  • Availability
  • Add to Cart