MAT196007274 Matrix Scientific

2-Bromo-N-(2-methylphenyl)propanamide

  • MDL number MFCD08670216
  • Cas Number : 19397-79-6
  • Molecular Weight : 242.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038467.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038467.html
Availability details for
Quantity
  • Availability
  • Add to Cart