MAT196007277 Matrix Scientific

2-Bromo-N-2,3-dihydro-1,4-benzodioxin-6-ylbutanamide

  • MDL number MFCD01994854
  • Cas Number : 347342-54-5
  • Molecular Weight : 300.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038470.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038470.html
Availability details for
Quantity
  • Availability
  • Add to Cart