MAT196007319 Matrix Scientific

2-{4-[3-(Trifluoromethyl)phenyl]piperazin-1-yl}ethanamine

  • MDL number MFCD10686739
  • Cas Number : 27144-85-0
  • Molecular Weight : 273.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038512.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038512.html
Availability details for
Quantity
  • Availability
  • Add to Cart