MAT196007328 Matrix Scientific

N-(2-Aminoethyl)-N-benzyl-N-ethylamine

  • MDL number MFCD03727288
  • Cas Number : 29950-34-3
  • Molecular Weight : 178.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/038521.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038521.html
Availability details for
Quantity
  • Availability
  • Add to Cart