MAT196007355 Matrix Scientific

1-(4-Methoxyphenyl)-2-methyl-1H-benzimidazole-5-carboxylic acid

  • MDL number MFCD02939955
  • Cas Number : 442531-47-7
  • Molecular Weight : 282.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038548.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038548.html
Availability details for
Quantity
  • Availability
  • Add to Cart