MAT196007368 Matrix Scientific

1-(3,5-Dimethylphenyl)-1H-benzimidazole-5-carboxylic acid

  • MDL number MFCD03021469
  • Cas Number : 442531-63-7
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038561.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038561.html
Availability details for
Quantity
  • Availability
  • Add to Cart