MAT196007402 Matrix Scientific

2-(1-Adamantyl)pyrrolidine

  • MDL number MFCD02663498
  • Cas Number : 180258-96-2
  • Molecular Weight : 205.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038595.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038595.html
Availability details for
Quantity
  • Availability
  • Add to Cart