MAT196007403 Matrix Scientific

5-Aminoadamantan-2-ol

  • MDL number MFCD09035852
  • Cas Number : 22591-69-1
  • Molecular Weight : 167.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248-250
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/038596.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038596.html
Availability details for
Quantity
  • Availability
  • Add to Cart