MAT196007414 Matrix Scientific

Ethyl 2-hydrazino-4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD01859678
  • Cas Number : 52481-66-0
  • Molecular Weight : 201.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038607.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038607.html
Availability details for
Quantity
  • Availability
  • Add to Cart