MAT196007436 Matrix Scientific

5-Chloro-2-(ethylthio)pyrimidine-4-carboxylic acid

  • MDL number MFCD00507248
  • Cas Number : 382610-58-4
  • Molecular Weight : 218.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/038629.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038629.html
Availability details for
Quantity
  • Availability
  • Add to Cart