MAT196007468 Matrix Scientific

3-(4-Bromophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD03834508
  • Cas Number : 618383-49-6
  • Molecular Weight : 267.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038661.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038661.html
Availability details for
Quantity
  • Availability
  • Add to Cart