MAT196007516 Matrix Scientific

1-Chloro-2-fluoro-3-isocyanatobenzene

  • MDL number MFCD03701597
  • Cas Number : 69922-25-4
  • Molecular Weight : 171.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/038709.html

Safety Information

hcodes Irritant/Lachrymator
MSDS http://www.matrixscientific.com/038709.html
Availability details for
Quantity
  • Availability
  • Add to Cart