MAT196007517 Matrix Scientific

Methyl 3-isocyanato-3-phenylpropanoate

  • MDL number MFCD04117485
  • Cas Number : 51031-37-9
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 152-153� Torr
Product Link http://www.matrixscientific.com/038710.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038710.html
Availability details for
Quantity
  • Availability
  • Add to Cart