MAT196007642 Matrix Scientific

1-Chloro-4-phenyl-5H-pyridazino[4,5-b]indole

  • MDL number MFCD12027475
  • Cas Number : 83749-78-4
  • Molecular Weight : 279.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038835.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038835.html
Availability details for
Quantity
  • Availability
  • Add to Cart