MAT196007738 Matrix Scientific

N-(1H-Pyrrol-2-ylmethyl)cyclopropanamine

  • MDL number MFCD12027502
  • Cas Number : 643008-01-9
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-120
Country of Origin USA
Product Link http://www.matrixscientific.com/038931.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038931.html
Availability details for
Quantity
  • Availability
  • Add to Cart