MAT196007848 Matrix Scientific

2,2-Dimethyl-3-(piperidin-1-ylcarbonyl)-cyclopropanecarboxylic acid

  • MDL number MFCD12027546
  • Cas Number : 1142215-00-6
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039041.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039041.html
Availability details for
Quantity
  • Availability
  • Add to Cart