MAT196007858 Matrix Scientific

3-[(2,6-Dimethylmorpholin-4-yl)carbonyl]-2,2-dimethylcyclopropanecarboxylic acid

  • MDL number MFCD12027556
  • Cas Number : 1186647-51-7
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039051.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039051.html
Availability details for
Quantity
  • Availability
  • Add to Cart