MAT196007860 Matrix Scientific

2,2-Dimethyl-3-[(3-methylpiperidin-1-yl)carbonyl]-cyclopropanecarboxylic acid

  • MDL number MFCD12027558
  • Cas Number : 1186654-05-6
  • Molecular Weight : 239.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039053.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039053.html
Availability details for
Quantity
  • Availability
  • Add to Cart