MAT196008145 Matrix Scientific

(4E)-3-(Chloromethyl)-4-(1H-indol-3-ylmethylene)-isoxazol-5(4H)-one

  • MDL number MFCD12027815
  • Cas Number : 1142199-56-1
  • Molecular Weight : 260.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039328.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039328.html
Availability details for
Quantity
  • Availability
  • Add to Cart