MAT196008315 Matrix Scientific

1H-Benzimidazole-6-carboxylic acid

  • MDL number MFCD03225617
  • Cas Number : 15788-16-6
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039498.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039498.html
Availability details for
Quantity
  • Availability
  • Add to Cart