MAT196008339 Matrix Scientific

[(5-Methyl-2-phenyl-1,3-oxazol-4-yl)methyl]amine

  • MDL number MFCD08271882
  • Cas Number : 132451-28-6
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039522.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039522.html
Availability details for
Quantity
  • Availability
  • Add to Cart