MAT196008399 Matrix Scientific

(5E)-5-(3-Chloro-4-hydroxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

  • MDL number MFCD12028042
  • Cas Number : 497835-71-9
  • Molecular Weight : 330.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039582.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039582.html
Availability details for
Quantity
  • Availability
  • Add to Cart