MAT196008478 Matrix Scientific

3-(4-Benzylpiperidin-1-yl)-3-oxopropanenitrile

  • MDL number MFCD01353601
  • Cas Number : 551907-33-6
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039661.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039661.html
Availability details for
Quantity
  • Availability
  • Add to Cart