MAT196008522 Matrix Scientific

2-Cyano-N-(2-nitrophenyl)acetamide

  • MDL number MFCD01910047
  • Cas Number : 65372-22-7
  • Molecular Weight : 205.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/039705.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039705.html
Availability details for
Quantity
  • Availability
  • Add to Cart