MAT196008554 Matrix Scientific

(1-Methyl-1H-pyrazol-3-yl)acetic acid

  • MDL number MFCD12028051
  • Cas Number : 1177318-00-1
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039737.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039737.html
Availability details for
Quantity
  • Availability
  • Add to Cart