MAT196008556 Matrix Scientific

2-(3-Chlorophenoxy)nicotinamide

  • MDL number MFCD02325548
  • Cas Number : 214758-92-6
  • Molecular Weight : 248.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039739.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039739.html
Availability details for
Quantity
  • Availability
  • Add to Cart