MAT196008573 Matrix Scientific

5-Amino-1-benzyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD02219534
  • Cas Number : 25784-56-9
  • Molecular Weight : 218.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/039756.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039756.html
Availability details for
Quantity
  • Availability
  • Add to Cart