MAT196008658 Matrix Scientific

2-Chloro-4-nitro-1H-imidazole

  • MDL number MFCD03419295
  • Cas Number : 57531-37-0
  • Molecular Weight : 147.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/039841.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039841.html
Availability details for
Quantity
  • Availability
  • Add to Cart