MAT196008679 Matrix Scientific

Pentane-2,3,4-trione 3-oxime

  • MDL number MFCD00466577
  • Cas Number : 29917-12-2
  • Molecular Weight : 129.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039862.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039862.html
Availability details for
Quantity
  • Availability
  • Add to Cart