MAT196008776 Matrix Scientific

5-(2-Aminoethyl)pyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD08449536
  • Cas Number : 221170-25-8
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/039959.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/039959.html
Availability details for
Quantity
  • Availability
  • Add to Cart