MAT196008823 Matrix Scientific

2-[(2-Fluorobenzyl)oxy]benzoic acid

  • MDL number MFCD09712403
  • Cas Number : 938279-93-7
  • Molecular Weight : 246.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040006.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040006.html
Availability details for
Quantity
  • Availability
  • Add to Cart