MAT196008825 Matrix Scientific

3-[(2-Chlorobenzyl)oxy]benzoic acid

  • MDL number MFCD04164523
  • Cas Number : 223102-78-1
  • Molecular Weight : 262.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/040008.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040008.html
Availability details for
Quantity
  • Availability
  • Add to Cart