MAT196008830 Matrix Scientific

2-Ethoxy-3-methoxybenzoic acid

  • MDL number MFCD06203281
  • Cas Number : 773869-96-8
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-52
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/040013.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040013.html
Availability details for
Quantity
  • Availability
  • Add to Cart