MAT196008875 Matrix Scientific

4-[(4-Methylbenzyl)oxy]benzenecarboxylic acid

  • MDL number MFCD00625986
  • Cas Number : 56442-19-4
  • Molecular Weight : 242.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040058.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040058.html
Availability details for
Quantity
  • Availability
  • Add to Cart