MAT196008940 Matrix Scientific

3-[(2,4-Dichlorobenzyl)oxy]benzoic acid

  • MDL number MFCD04537474
  • Cas Number : 360778-55-8
  • Molecular Weight : 297.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040123.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040123.html
Availability details for
Quantity
  • Availability
  • Add to Cart