MAT196008951 Matrix Scientific

2-{[3-(Trifluoromethyl)benzyl]oxy}benzoic acid

  • MDL number MFCD12028133
  • Cas Number : 1040038-41-2
  • Molecular Weight : 296.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/040134.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040134.html
Availability details for
Quantity
  • Availability
  • Add to Cart