MAT196008987 Matrix Scientific

(5E)-5-(4-Chlorobenzylidene)-2-mercapto-1,3-thiazol-4(5H)-one

  • MDL number MFCD04969011
  • Cas Number : 81154-18-9
  • Molecular Weight : 255.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/040170.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040170.html
Availability details for
Quantity
  • Availability
  • Add to Cart