MAT196009035 Matrix Scientific

1-Ethyl-5-methyl-1H-pyrazole-4-carbonyl chloride

  • MDL number MFCD03421498
  • Cas Number : 1172423-27-6
  • Molecular Weight : 172.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040218.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040218.html
Availability details for
Quantity
  • Availability
  • Add to Cart