MAT196009238 Matrix Scientific

[(3S,4R)-4-(2-Methoxyphenyl)pyrrolidin-3-yl]-methanol hydrochloride

  • MDL number MFCD12028176
  • Cas Number : 1217660-06-4
  • Molecular Weight : 243.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040302.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040302.html
Availability details for
Quantity
  • Availability
  • Add to Cart