MAT196009247 Matrix Scientific

[(3S,4R)-4-Methyl-4-phenylpyrrolidin-3-yl]-methanol hydrochloride

  • MDL number MFCD12028190
  • Cas Number : 1217842-07-3
  • Molecular Weight : 227.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040316.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040316.html
Availability details for
Quantity
  • Availability
  • Add to Cart