MAT196009275 Matrix Scientific

{2-[1-(Phenylsulfonyl)piperidin-4-yl]ethyl}aminehydrochloride

  • MDL number MFCD12028330
  • Cas Number : 1160245-35-1
  • Molecular Weight : 304.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/040624.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040624.html
Availability details for
Quantity
  • Availability
  • Add to Cart