MAT196009291 Matrix Scientific

{2-[3-(3-Methylbenzyl)-1H-1,2,4-triazol-5-yl]-ethyl}amine hydrochloride

  • MDL number MFCD12028351
  • Cas Number : 1185297-97-5
  • Molecular Weight : 252.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040645.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040645.html
Availability details for
Quantity
  • Availability
  • Add to Cart