MAT196009299 Matrix Scientific

4-Benzylpiperidine-4-carboxylic acid hydrochloride

  • MDL number MFCD12028367
  • Cas Number : 1184995-85-4
  • Molecular Weight : 255.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040661.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040661.html
Availability details for
Quantity
  • Availability
  • Add to Cart