MAT196009305 Matrix Scientific

[(3-Ethyl-4,5-dihydroisoxazol-5-yl)methyl]aminehydrochloride

  • MDL number MFCD12028412
  • Cas Number : 1160245-60-2
  • Molecular Weight : 164.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040827.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040827.html
Availability details for
Quantity
  • Availability
  • Add to Cart