MAT196009344 Matrix Scientific

Benzyl-methyl-piperidin-3-yl-amine hydrochloride

  • MDL number MFCD12028485
  • Cas Number : 1185296-10-9
  • Molecular Weight : 240.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041034.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041034.html
Availability details for
Quantity
  • Availability
  • Add to Cart