MAT196009352 Matrix Scientific

4-(2-Phenoxy-ethyl)-morpholine-2-carboxylic acid

  • MDL number MFCD12028493
  • Cas Number : 1171916-86-1
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041044.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041044.html
Availability details for
Quantity
  • Availability
  • Add to Cart