MAT196009369 Matrix Scientific

2-(1-Benzyl-piperidin-2-yl)-ethylaminedihydrobromide

  • MDL number MFCD12028511
  • Cas Number : 1185079-12-2
  • Molecular Weight : 380.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041064.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041064.html
Availability details for
Quantity
  • Availability
  • Add to Cart