MAT196009385 Matrix Scientific

3-Piperidin-4-yl-1H-pyrazolo[3,4-b]pyrazinedihydrochloride

  • MDL number MFCD12028526
  • Cas Number : 1185112-92-8
  • Molecular Weight : 276.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041081.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041081.html
Availability details for
Quantity
  • Availability
  • Add to Cart